Learn More
Invitrogen™ WNT10B Polyclonal Antibody

Description
A suggested positive control is human skeletal muscle tissue lysate. PA5-20530 can be used with blocking peptide PEP-0650.
Specifications
Specifications
| Antigen | WNT10B |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | WNT10B |
| Gene Accession No. | O00744, P48614 |
| Gene Alias | Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12 |
| Gene Symbols | WNT10B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.