Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547362
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547362 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547362 Supplier Invitrogen™ Supplier No. PA547362
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) Wnt-3A is observed and less than 1% cross-reactivity with rmWnt-5A is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT11
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene WNT11
Gene Accession No. O96014, P48615
Gene Alias HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11
Gene Symbols WNT11
Host Species Goat
Immunogen E. coli-derived recombinant mouse Wnt-11 peptide Leu39-Ala79 and Lys225-Arg297.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22411, 7481
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.