Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111305 Supplier Invitrogen™ Supplier No. PA5111305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKTPSALALN QTQHCKQLEG LVSAQVQLCR SNLELMHTVV HAAREVMKAC RRAFADMRWN CSSIELAPNY LLDLERGTRE Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 94%.

Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WNT11
Gene Accession No. O96014
Gene Alias HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11
Gene Symbols WNT11
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human WNT11. Recombinant protein control fragment (Product #RP-101129).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7481
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.