Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt16 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299544AF532
Description
Wnt16 Polyclonal specifically detects Wnt16 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Wnt16 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
protein Wnt-16, wingless-type MMTV integration site family, member 16 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse Wnt16. | |
0.1 mL | |
Wnt Signaling Pathway | |
51384 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction