Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt16 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299324MFV450
Description
Wnt16 Polyclonal antibody specifically detects Wnt16 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Wnt16 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
protein Wnt-16, wingless-type MMTV integration site family, member 16 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Wnt16. | |
0.1 mL | |
Wnt Signaling Pathway | |
51384 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction