Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT2 Mouse anti-Human, Mouse, Rat, Clone: 3H7B10, Proteintech
SDP

Catalog No. 501737074 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7074 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7074 Supplier Proteintech Group Inc Supplier No. 666561IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.

Specifications

Antigen WNT2
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H7B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene WNT2
Gene Accession No. P09544, P21552
Gene Alias Int 1 like protein 1, Int 1 related protein, INT1L1, IRP, Protein Wnt 2, WNT2
Gene Symbols WNT2
Host Species Mouse
Immunogen WNT2 Fusion Protein Ag25185
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114487, 22413, 7472
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.