Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596453 Supplier Invitrogen™ Supplier No. PA596453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SWWYMRATGG SSRVMCDNVP GLVSSQRQLC HRHPDVMRAI SQGVAEWTAE CQHQFRQHRW NCNTLDRDHS LFGRVLLRSS RESAFVYAIS SAGVVFAITR ACSQGEVKSC SCDPKKMGSA KDSKGIFDWG GCSDNIDYGI KFARAFVDAK ERKGKDARAL MNLHNNRAGR KAVKRFLKQE CKCHGVSGSC TLRTCWLAMA DFRKTGDYLW RKYNGAIQVV MNQDGTGFTV ANERFKKPTK NDLVYFENSP DYCIRDREAG SLGTAGRVCN LTSRGMDSCE VMCCGRGYDT SHVTRMTKCG CKFHWCCAVR CQDCLEALDV HTCKAPKNAD WTTAT; Positive Samples: SKOV3; Cellular Location: extracellular matrix, extracellular space.

WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Wnt2
Gene Accession No. P09544
Gene Alias 2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2
Gene Symbols Wnt2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-360 of human WNT2 (NP_0033821).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114487, 7472
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.