Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT2 Recombinant Rabbit Monoclonal Antibody (HL1999)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550358
Description
WNT2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Specifications
WNT2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P09544, P21552 | |
Wnt2 | |
Recombinant fragment of human Wnt2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
HL1999 | |
Unconjugated | |
Wnt2 | |
2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2 | |
Rabbit | |
Protein A | |
RUO | |
114487, 22413, 7472 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction