Learn More
Invitrogen™ WNT2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533113
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Hamster, Panda, Horse, Rabbit, Pig (100%); Elephant (94%); Opossum, Platypus (88%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
WNT2B is a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. It may play a role in human development as well as human carcinogenesis. Two alternatively spliced transcript variants exist.
Specifications
| WNT2B | |
| Polyclonal | |
| Unconjugated | |
| WNT2B | |
| Protein Wnt-13; Protein Wnt-2b; wingless related MMTV integration site 2b; wingless-type MMTV integration site 2B; wingless-type MMTV integration site family member 2B; wingless-type MMTV integration site family, member 13; wingless-type MMTV integration site family, member 2B; Wnt family member 2B; Wnt13; wnt-13; Wnt2b; XWNT2, Xenopus, homolog of | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100063931, 100349847, 100520560, 100761413, 116466, 22414, 445420, 483220, 7482 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O70283, Q93097 | |
| WNT2B | |
| Synthetic 16 amino acid peptide from internal region of human WNT2B. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.