Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585505 Supplier Invitrogen™ Supplier No. PA585505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Chicken (80%), Bovine (99%). Positive Control: HepG2 conditioned medium. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

WNT2B is a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. It may play a role in human development as well as human carcinogenesis. Two alternatively spliced transcript variants exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT2B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene WNT2B
Gene Accession No. Q93097
Gene Alias Protein Wnt-13; Protein Wnt-2b; wingless related MMTV integration site 2b; wingless-type MMTV integration site 2B; wingless-type MMTV integration site family member 2B; wingless-type MMTV integration site family, member 13; wingless-type MMTV integration site family, member 2B; Wnt family member 2B; Wnt13; wnt-13; Wnt2b; XWNT2, Xenopus, homolog of
Gene Symbols WNT2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WNT2B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.