Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT2B Recombinant Rabbit Monoclonal Antibody (JB48-34)
GREENER_CHOICE

Catalog No. PIMA542480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542480 Supplier Invitrogen™ Supplier No. MA542480
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: SiHa, Hela, LOVO, SHSY-5Y, human kidney tissue, mouse kidney tissue, rat testis tissue. Subcellular Location: Secreted. Extracellular region.

WNT2B is a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. It may play a role in human development as well as human carcinogenesis. Two alternatively spliced transcript variants exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT2B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB48-34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene WNT2B
Gene Accession No. O70283, Q93097
Gene Alias Protein Wnt-13; Protein Wnt-2b; wingless related MMTV integration site 2b; wingless-type MMTV integration site 2B; wingless-type MMTV integration site family member 2B; wingless-type MMTV integration site family, member 13; wingless-type MMTV integration site family, member 2B; Wnt family member 2B; Wnt13; wnt-13; Wnt2b; XWNT2, Xenopus, homolog of
Gene Symbols WNT2B
Host Species Rabbit
Immunogen Recombinant protein within Human Wnt2b aa 175-355 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116466, 22414, 7482
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.