Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt3 Antibody (OTI1G7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274869IR
Description
Wnt3 Monoclonal specifically detects Wnt3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt3 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WNT3 | |
Recombinant protein fragment corresponding to amino acids 128-355 of human Wnt3 (NP_110380) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1G7 | |
Western Blot, Immunohistochemistry | |
INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein | |
Mouse | |
Affinity Purified | |
Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7473.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction