Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578226 Supplier Invitrogen™ Supplier No. PA578226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Wnt3-transfected 293T cells, Want3A-transfected 293T cells Predicted Reactivity: Mouse (99%), Rat (99%), Zebrafish (91%), Chicken (98%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

WNT3, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT3
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene WNT3
Gene Accession No. P56703
Gene Alias INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein
Gene Symbols WNT3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WNT3. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7473
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.