Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580230
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580230 Supplier Invitrogen™ Supplier No. PA580230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Human Placenta Tissue, HEPG2 whole cell.

WNT3, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT3
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene WNT3
Gene Accession No. P56703
Gene Alias INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein
Gene Symbols WNT3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Wnt3 (185-207aa AMNKHNNEAGRTTILDHMHLKCK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7473
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.