Learn More
Invitrogen™ WNT3A Polyclonal Antibody

Description
Positive Samples: U2OS, A-549, PC-3, Raji, 293T, Mouse lung, Mouse testis, Rat lung, Rat testis; Cellular Location: extracellular matrix, extracellular space Immunogen sequence: SSRHQGSPGK GWKWGGC.
Specifications
Specifications
| Antigen | WNT3A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | WNT3A |
| Gene Accession No. | P56704 |
| Gene Alias | MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a] |
| Gene Symbols | WNT3A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.