Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech WNT3A Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 502718568
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8568 500 μg
500P251BT25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-271-8568 Supplier PeproTech Supplier No. 500P251BT500UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-3a. Anti-Human Wnt-3a-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hWNT-3a by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human WNT-3a (500-P251) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hWNT-3a. Western Blot: To detect hWNT-3a by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hWNT-3a is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P251BT-1MG will be provided as 2 x 500 μg

Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT3A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene WNT3A
Gene Accession No. P56704
Gene Alias MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a]
Gene Symbols WNT3A
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human Wnt-3a.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89780
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.