Learn More
WNT4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The WNT4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNT4. This antibody recognizes human antigen. The WNT4 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
| Antigen | WNT4 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Concentration | 267 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 54361 |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.