Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT4 Recombinant Rabbit Monoclonal Antibody (HL1889)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550344
Description
WNT4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family, and is the first signaling molecule shown to influence the sex-determination cascade. It encodes a protein which shows 98% amino acid identity to the Wnt4 protein of mouse and rat. This gene and a nuclear receptor known to antagonize the testis-determining factor play a concerted role in both the control of female development and the prevention of testes formation. This gene and another two family members, WNT2 and WNT7B, may be associated with abnormal proliferation in breast tissue. Mutations in this gene can result in Rokitansky-Kuster-Hauser syndrome and in SERKAL syndrome.
Specifications
WNT4 | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with no preservative | |
P56705 | |
Wnt4 | |
Recombinant fragment of human Wnt4. | |
100 μL | |
Primary | |
Human, Zebrafish | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry, Western Blot | |
HL1889 | |
Unconjugated | |
Wnt4 | |
protein Wnt-4; SERKAL; signal molecule; UNQ426/PRO864; wingless-related MMTV integration site 4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4; Wnt family member 4; WNT4; Wnt-4 | |
Rabbit | |
Protein A | |
RUO | |
54361 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction