Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT5A Monoclonal Antibody (C3)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | WNT5A |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541694
|
Invitrogen™
MA541694 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543545
|
Invitrogen™
MA543545 |
20 μL |
Each for $243.00
|
|
|||||
Description
WNT5A Monoclonal Antibody for Western Blot
Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.Specifications
WNT5A | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P22725 | |
22418 | |
Recombinant protein Wingless Type MMTV Integration Site Family, Member 5A. The antigen corresponds to amino acid range 62-380 of the target protein. | |
Antibody |
C3 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Mouse | |
WNT5A | |
8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein | |
WNT5A | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title