Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120790 Supplier Invitrogen™ Supplier No. PA5120790
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SKOV3, HeLa, Mouse lung, Mouse kidney, Mouse brain. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: AQPLCSQLAG LSQGQKKLCH LYQDHMQYIG EGAKTGIKEC QYQFRHRRWN CSTVDNTSVF GRVMQIGSRE TAFTYAVSAA GVVNAMSRAC REGELSTCGC SRAARPKDLP RDWLWGGCGD NIDYGYRFAK EFVDARERER IHAKGSYESA RILMNLHNNE AGRRTVYNLA DVACKCHGVS GSCSLKTCWL QLADFRKVGD ALKEKYDSAA AMRLNSRGKL VQVNSRFNSP TTQDLVYIDP S.

Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT5A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WNT5A
Gene Accession No. P22725, P41221, Q9QXQ7
Gene Alias 8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein
Gene Symbols WNT5A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 65-305 of human WNT5A (NP_0033832).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22418, 64566, 7474
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.