Learn More
Invitrogen™ WNT5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533115
Description
Percent identity with other species by BLAST analysis: Human, Orangutan, Gibbon, Monkey (100%); Gorilla, Marmoset (93%); Dog, Elephant, Horse, Rabbit, Opossum (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants.
Specifications
WNT5B | |
Polyclonal | |
Unconjugated | |
WNT5B | |
AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81029 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H1J7 | |
WNT5B | |
Synthetic 14 amino acid peptide from internal region of human WNT5B. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.