Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT5B Recombinant Rabbit Monoclonal Antibody (JE53-67)
GREENER_CHOICE

Catalog No. PIMA536157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536157 Supplier Invitrogen™ Supplier No. MA536157
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

WNT5B Recombinant Monoclonal Antibody for Flow, Western Blot

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT5B
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone JE53-67
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene WNT5B
Gene Accession No. Q9H1J7
Gene Alias AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein
Gene Symbols WNT5B
Host Species Rabbit
Immunogen Recombinant protein within Human WNT5B aa 260-359.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81029
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.