Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534326
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534326 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534326 Supplier Invitrogen™ Supplier No. PA534326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Dog, Horse, Rabbit (100%).

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene WNT6
Gene Accession No. P22727, Q9Y6F9
Gene Alias AA409270; Protein Wnt-6; wingless-related MMTV integration site 6; wingless-related MMTV integration site 6 homolog; wingless-type MMTV integration site family member 6; wingless-type MMTV integration site family, member 6; Wnt family member 6; WNT6; Wnt-6
Gene Symbols WNT6
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from N-terminus of human WNT6.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100058824, 100348986, 100751289, 22420, 316526, 488527, 505075, 7475
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Hamster, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.