Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578227 Supplier Invitrogen™ Supplier No. PA578227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: V5-human Wnt7A-transfected 293T cells Predicted Reactivity: Mouse (100%), Rat (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Wnt-7a belongs to the Wnt family of signaling proteins that play a key role in maintaining the integrity of embryonic and adult tissues. It is expressed in placenta, kidney, testis, uterus, fetal lung, and fetal and adult brain. Most Wnt proteins can signal though a mechanism called the canonical Wnt pathway, in which Wnt proteins bind to and activate seven-pass transmembrane receptors of the Frizzled family, ultimately leading to the disruption of β-catenin degradation. Intracellular accumulation of β-catenin increases translocation of the protein into the nucleus, where it binds to TCF/LEF transcription factors to induce the expression of numerous genes. Increased Wnt/β-catenin signaling is associated with tumorigenesis in a diverse set of human cancers. However, Wnt-7a/Frizzled-9 signaling has been shown to act as a tumor suppressor in non-small cell lung cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT7A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Wnt7a
Gene Accession No. O00755
Gene Alias AI849442; postaxial hemimelia; protein Wnt-7a; proto-oncogene Wnt7a protein; px; tw; wingless-related MMTV integration site 7A; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family, member 7A; Wnt family member 7A; WNT7A; Wnt-7a
Gene Symbols Wnt7a
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WNT7A. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114850, 7476
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.