Learn More
Invitrogen™ WNT7A Polyclonal Antibody

Description
Recommended positive controls: WNT7A-transfected 293T. Predicted reactivity: Mouse (100%), Rat (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | WNT7A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Wnt7a |
| Gene Accession No. | O00755, P24383 |
| Gene Alias | AI849442; postaxial hemimelia; protein Wnt-7a; proto-oncogene Wnt7a protein; px; tw; wingless-related MMTV integration site 7A; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family, member 7A; Wnt family member 7A; WNT7A; Wnt-7a |
| Gene Symbols | Wnt7a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.