Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT7B Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody

$243.00 - $552.50

Specifications

Antigen WNT7B
Clone C4
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543544
View Documents
Invitrogen™
MA543544
20 μL
Each for $243.00
Only null left
Add to Cart
 
PIMA543543
View Documents
Invitrogen™
MA543543
100 μL
Each for $552.50
Only null left
Add to Cart
 
Description

Description

Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.
Specifications

Specifications

WNT7B
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300
Protein Wnt-7b; wingless-related MMTV integration site 7B; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7B; Wnt family member 7B; WNT7B; Wnt-7b
Wnt7b
IgG1 κ
Protein A/G
Antibody
C4
Western Blot
Unconjugated
Mouse
Human
P56706
7477
Thr216-Lys349 (Accession P56706), with Two N-terminal Tags, His-tag and SUMO-tag.
Primary
-20°C
Wnt7b
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.