Learn More
Invitrogen™ WNT7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110583
Description
Immunogen sequence: TAACSQGNLS NCGCDREKQG YYNQAEGWKW GGCSADVRYG IDFSRRFVDA REIKKNARRL MNLHNNEAGR KVLEDRMQLE CKCHGVSGSC TTKTCWTTLP KFREVGHLLK EKYNAAVQVE VVRASRLRQP TFLRIKQLRS YQKPMETDLV YIEKSPNYCE EDAATGSVGT QGRLCNRTSP GADGCDTMCC GRGYNTHQYT KVWQCNCKFH WCCFVKCNTC SERTEVFTCK.
Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.
Specifications
| WNT7B | |
| Polyclonal | |
| Unconjugated | |
| Wnt7b | |
| Protein Wnt-7b; wingless-related MMTV integration site 7B; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7B; Wnt family member 7B; WNT7B; Wnt-7b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7477 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56706 | |
| Wnt7b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-349 of human WNT7B (NP_478679.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.