Learn More
Invitrogen™ WNT8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533116
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bat, Elephant, Panda, Horse (100%); Bovine, Dog (93%); Rabbit (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
WNT8A, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
| WNT8A | |
| Polyclonal | |
| Unconjugated | |
| Wnt8a | |
| etID309727.14; protein Wnt-8a; protein Wnt-8a ORF1; protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; Stra11; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; Wnt family member 8A; wnt8; wnt-8; wnt8.1; Wnt8a; Wnt-8A; Wnt8d; Wnt-8D; Wnt8-like protein 1; wu:fa20e02; wu:fe05d07 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100072609, 7478 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9H1J5 | |
| Wnt8a | |
| Synthetic 14 amino acid peptide from internal region of human WNT8A. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Horse, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.