Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547278
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547278 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547278 Supplier Invitrogen™ Supplier No. PA547278
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3A, rmWnt-4, rmWnt-5A, and rmWnt-10B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

WNT8A, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT8A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Wnt8a
Gene Accession No. Q64527
Gene Alias etID309727.14; protein Wnt-8a; protein Wnt-8a ORF1; protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; Stra11; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; Wnt family member 8A; wnt8; wnt-8; wnt8.1; Wnt8a; Wnt-8A; Wnt8d; Wnt-8D; Wnt8-like protein 1; wu:fa20e02; wu:fe05d07
Gene Symbols Wnt8a
Host Species Goat
Immunogen E. coli-derived recombinant mouse Wnt-8a peptide Trp23-Glu53, Leu196-Arg273.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20890
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.