Learn More
Invitrogen™ WNT8B Polyclonal Antibody

Description
The antibody detects endogenous levels of total WNT8B / Wnt 8b protein.
Specifications
Specifications
| Antigen | WNT8B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WNT8B |
| Gene Accession No. | Q93098 |
| Gene Alias | protein Wnt-8b; wingless related MMTV integration site 8b; wingless-type MMTV integration site family member 8B; wingless-type MMTV integration site family, member 8B; WNT 8b; Wnt family member 8B; WNT8B; wnt-8b |
| Gene Symbols | WNT8B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.