Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572263
Description
WNT8B Polyclonal Antibody for Western Blot
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 95%, 86% and 71% amino acid identity to the mouse, zebrafish and Xenopus Wnt8B proteins, respectively. The expression patterns of the human and mouse genes appear identical and are restricted to the developing brain. The chromosomal location of this gene to 10q24 suggests it as a candidate gene for partial epilepsy.
Specifications
WNT8B | |
Polyclonal | |
Unconjugated | |
WNT8B | |
protein Wnt-8b; wingless related MMTV integration site 8b; wingless-type MMTV integration site family member 8B; wingless-type MMTV integration site family, member 8B; WNT 8b; Wnt family member 8B; WNT8B; wnt-8b | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
30144 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P51029 | |
WNT8B | |
KLH conjugated synthetic peptide between 201-235 amino acids from the central region of Zebrafish wnt8b. | |
200 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction