Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT9A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984309
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4309 Supplier Bioss Supplier No. BS1933R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WNT9A is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. WNT9A is likely to signal over only few cell diameters.

Specifications

Antigen WNT9A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene WNT9A
Gene Accession No. O14904
Gene Alias MGC138165; MGC141991; Protein Wnt-14; Protein Wnt-9a; wingless related MMTV integration site 9A; wingless-type MMTV integration site 14; wingless-type MMTV integration site 9A; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A; Wnt family member 9A; Wnt14; wnt-14; Wnt9a
Gene Symbols WNT9A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human WNT9A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7483
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.