Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534335
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534335 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534335 Supplier Invitrogen™ Supplier No. PA534335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Horse, Pig (100%) Bat, Rabbit (94%).

WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT9B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Wnt9b
Gene Accession No. O14905, O35468
Gene Alias clf; clf1; protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9B; Wnt family member 9B; Wnt14b; wnt-14b; Wnt15; wnt-15; Wnt9b
Gene Symbols Wnt9b
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human WNT9B.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100063730, 100738655, 100766088, 22412, 303586, 541243, 7484
Target Species Human, Mouse, Rat, Monkey, Bovine, Hamster, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.