Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT9B Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA548036
Description
In Western blots, less than 5% cross-reactivity with recombinant human (rh) Wnt-2, rhWnt-9a, recombinant mouse (rm) Wnt-1, rmWnt-2b, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-11, and rmWnt-16 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
WNT9B | |
Polyclonal | |
Unconjugated | |
Wnt9b | |
clf; clf1; protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9B; Wnt family member 9B; Wnt14b; wnt-14b; Wnt15; wnt-15; Wnt9b | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22412 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35468 | |
Wnt9b | |
E. coli-derived recombinant mouse Wnt-9b Ala46-Glu90, Lys228-Tyr298. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction