Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578231
Description
Positive Control: WNT9B-V5 transfected 293T lysates Predicted Reactivity: Mouse (100%), Rat (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
WNT9B | |
Polyclonal | |
Unconjugated | |
Wnt9b | |
clf; clf1; protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9B; Wnt family member 9B; Wnt14b; wnt-14b; Wnt15; wnt-15; Wnt9b | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.98 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O14905 | |
Wnt9b | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human WNT9B. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction