Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WNTinib
GREENER_CHOICESDP

Catalog No. 810650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-065-0 50 mg
81-061-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 81-065-0 Supplier Tocris Bioscience™ Supplier No. 8106/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of mutant β-catenin signaling in hepatocellular carcinoma

WNTinib is a multi-kinase inhibitor with selectivity for mutant β-catenin (CTNNB1) in hepatocellular carcinoma compared to wild type hepatocytes (IC50 ranges are 0.36 μM-0.54 μM in primary organoids; 0.28 μM-1 μM in primary human HCC cell lines and 0.45 μM-2.3 μM in human cell lines ). Also inhibits VEGFR2, PDE6D and VEGFR1 (IC50 values are 0.3 μM, 0.3 μM and 0.6 μM respectively). Downregulates WNT and phospho-EZH2, upregulates trimethylation of H3K27 and normalises expression of apoptotic genes.

Specifications

Chemical Name or Material 4-[3-Fluoro-4-[[[[4-(1,1,2,2,2-pentafluoroethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide
CAS 2770091-44-4
Quantity 50 mg
Target beta-Catenin
Molecular Formula C22H16F6N4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.