Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Women's Curvy Fit Flame Resistant Jean with Stretch

Catalog No. 17204252
Click to view available options
Size:
12
22
Size (English) Inseam:
30 in.
35 in.

Women's Curvy Fit Flame Resistant Jean with Stretch feel like an everyday jean. Light hand sanding and whiskering gives a more fashionable appearance. Felled inseam and seat seam adds durability. Contoured waistband provides a more comfortable fit.

  • Lightweight fabric with stretch that makes this work jean feel like an everyday jean.
  • Light hand sanding and whiskering gives a more fashionable appearance.
  • Felled inseam and seat seam adds durability.
  • Contoured waistband provides a more comfortable fit.
  • Metal button closure (metal is not exposed to the skin).
  • Two patch on spade shaped hip pockets.
  • Scoop front pockets with watch pocket on right side.
  • Zipper fly closure.
  • Certifications Level 1: NFPA 70E, NFPA 2112
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 5
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 12.5 oz. (425 g/m&superscript_2;) Flame Resistant Denim
  • Blend: Body: 98% Cotton / 2% Spandex Elastane
  • Waistband: Contoured waistband with stretch interlining
  • Features: Contoured waistband, Stretch fabric, Hand sanded fabric with whiskering, Felled inseam and seat seam
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Curvy
  • Gender: Female
  • Brand: Bulwark™

Specifications

Size 22
Gender Women's
Material Cotton, Polyester, Spandex, Polyester, Spandex
Color Denim
Seam Type Stitched
No. of Pockets 4
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 21
Certifications/Compliance NFPA 70E, NFPA 2112
Size (English) Inseam 35 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.