Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Women's Curvy Fit Flame Resistant Jean with Stretch
Click to view available options
Size:
0
2
4
6
8
10
12
14
16
18
20
22
24
Size (English) Inseam:
30 in.
32 in.
35 in.
Description
- Lightweight fabric with stretch that makes this work jean feel like an everyday jean.
- Light hand sanding and whiskering gives a more fashionable appearance.
- Felled inseam and seat seam adds durability.
- Contoured waistband provides a more comfortable fit.
- Metal button closure (metal is not exposed to the skin).
- Two patch on spade shaped hip pockets.
- Scoop front pockets with watch pocket on right side.
- Zipper fly closure.
- Certifications Level 1: NFPA 70E, NFPA 2112
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 5
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 12.5 oz. (425 g/m&superscript_2;) Flame Resistant Denim
- Blend: Body: 98% Cotton / 2% Spandex Elastane
- Waistband: Contoured waistband with stretch interlining
- Features: Contoured waistband, Stretch fabric, Hand sanded fabric with whiskering, Felled inseam and seat seam
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Curvy
- Gender: Female
- Brand: Bulwark™
Specifications
Specifications
Size | 0 |
Gender | Women's |
Material | Cotton, Polyester, Spandex, Polyester, Spandex |
Color | Denim |
Seam Type | Stitched |
No. of Pockets | 4 |
Closure Type | Concealed Button |
Disposable | Reusable |
Hazard Risk Category | Category 2 |
Arc Flash Rating | 21 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction