Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wood's metal, rods

Catalog No. p-3593544
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 76093-98-6 | Pb·Sn

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76093-98-6
Molecular Formula Pb·Sn
MDL Number MFCD00198175

Specifications

Color Silver to Gray
Melting Point 70.0°C
Quantity 100 g
Packaging Glass bottle
Form Rod
Formula Weight 647.28
Chemical Name or Material Wood's metal
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid contact during pregnancy/while nursing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.