Learn More
MSA™ Workman™ FP Stryder™ Beam Grip
Workman FP Stryder™ Beam Anchors quickly and easily attach to an I-beam flange without the need for any tools.
$394.12 - $594.52
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-166-582
View Documents
MSA™
10144431
4 - 13.5 in. beam
Each for $394.12
Only null left
Add to Cart
19-166-583
View Documents
MSA™
10144432
14 - 23.5 in. beam
Each for $594.52
Only null left
Add to Cart
Description
Description
  • Compatible for use with MSA fall arrest and confined space entry/retrieval components
  • Attaches to I-beam flange - no tools required
Compliance
Meets applicable OSHA standards; ANSI™ Z359.1-2007 requirements.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.