Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Workman™ Full Body Harnesses

Catalog No. 19825019 Shop All MSA Products
Change view
Click to view available options
Buckle Style (Leg):
Tongue leg buckles
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) Size
19-825-019 Tongue leg buckles Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-825-019 Supplier MSA™ Supplier No. 10072487

For working in confined spaces and at heights. MSA™ Workman™ Harnesses provide optimum quality and fall arrest systems for workers.

  • Provides protection due to its lightweight components and durable webbing
  • Vest-style harness with optional hip D-Rings, fits wide range of body types and sizes
  • Color-contrasting thigh and torso straps simplify donning
  • Econo

Certifications

CSA Z259.10-07

Compliance

OSHA regulations, ANSI A10.32, ANSI Z359.2

Specifications

Color Yellow
Material Stainless Steel
Webbing Material Polyester
Product Type Full body harness
Size Standard
For Use With (Application) Working in confined spaces and at heights
D-Ring Location Back
Buckle Style (Leg) Tongue leg buckles
Certifications/Compliance OSHA regulations, ANSI A10.32, ANSI Z359.1, CSA Z259.10-06
Buckle Style (Chest) Qwik-Fit chest strap

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.