Learn More
Bulwark™ CP Chemical Splash Protection Lab Coats
Catalog No. 19823610 Shop All Red Kap Industries Products
This item is not returnable. View return policy
Catalog No. 19-823-610 Supplier Bulwark™ CP Supplier No. 352CWH XL 0R
This item is not returnable. View return policy
Bulwark™ CP Chemical Splash Protection Lab Coatc with Milliken ShieldCSR chemical splash protection finish that resists penetration and repels inadvertent liquid chemical splash.
  • Featuring Milliken ShieldCSR Chemical Splash protection finish resists penetration and repels inadvertent liquid chemical splash
  • Snap front closure for easy on/off
  • Lay-down collar
  • Knit rib cuffs for better fit at wrist
  • Splash resistant side pass-throughs for easy inner pocket access
  • Pockets include one chest pocket on left chest with a pencil slot and two large lower front pockets with flaps
Specifications
Product Type Chemical Resistant Lab Coat
Gender Unisex
Color White
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals