Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR FR/CP Industrial Coverall

Catalog No. 19823628 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-628 Large
19-823-624 Small
19-823-625 Small
19-823-626 Medium
19-823-627 Medium
19-823-629 Large
19-823-630 X-Large
19-823-631 X-Large
19-823-632 2X-Large
19-823-633 2X-Large
19-823-634 3X-Large
19-823-635 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-823-628 Supplier Bulwark™ FR Supplier No. 1064RBLGLN
This item is not returnable. View return policy

Bulwark™ FR FR/CP Industrial Coverall Nomex™ IIIA with ShieldCXP provides flame resistant and chemical splash protection.

  • Stand up collar
  • Action back for ease of movement
  • Pass-through access to inner clothing
  • Solid brass breakaway zipper
  • Snap wrist closures
  • Roomy leg cut to fit over boots
  • Pleated elastic waist front and back for superior fit
  • Pockets include two extra-deep front inset pockets, two rear patch pockets, two large double-stitched chest pockets with snap flap closure
  • NFPA 2112 UL-Certifieds

Specifications

Product Type FR/CP Coveralls
Size Large
Gender Unisex
Material Dupont™ Nomex™
Color Blue
Disposable Reusable
Waist Style Pleated Elastic
Ankle Style Open
Cuff Style Snap
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 1
Resistance Chemicals
Coating Westex ShieldCXP™
Collar Type Standing
Arc Flash Rating ATPV 5.6
Certifications/Compliance NFPA 2112
Description Flame-resistant and chemical-splash protection coveralls
Latex Free Yes
No. of Pockets 6
Size (English) Large Long
For Use With (Application) Arc Flash and Chemical Splash
Quantity 1/Ea.
Style Long
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.