Learn More
Brady™ Workstation Scan and Print Suite CD
Catalog No. 19129126 Shop All Brady Corporation Products
Click to view available options
Quantity:
1 Ea.
Catalog No. 19-129-126
Supplier: Brady™ BWSSPSCD
Brady Workstation Scan and Print Suite CD, create production label workflows that you can pair with a barcode scanning device and Brady printer.
Scan Mode - Scan existing barcodes to capture data and transfer onto a new label template.CVS Mode - Scan the name of a CSV file to print a specified batch of labelsCustom Script Mode - Perform advanced label operations such as parse scanned data or serialize labelsScalable Workflows - Expand workflows to additional workstations on large projects using the Export and Import feature Quickly replicate labels in a lab for sample testing, for aliquots at a bio bank, or in a variety of other environments
Specifications
Product Type Printer
Display Type None
For Use With (Equipment) Brady Labels, Brady Scanners
Material Plastic
Width (English) 7.6 in.
Width (Metric) N/A
Includes CD with software
Operating System Windows™ 7 (32- or 64-bit);Windows™ 7 Enterprise;Windows™ 7 Professional;Windows™ 7 SPI;Windows™ 7 Ultimate;Windows™ 8 (32- or 64-bit);Windows™ 8 Professional (32 or 64 bit);Windows™ 8.1 (32- or 64-bit);Windows™ 8.1 Enterprise;Windows™ 8.1 Professional;Windows™ 10
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.