Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Worm Composting Kit and Video

Click to view available options
Product Type:
Redworms Coupon
Wormania! Video to Compliment Worm-A-Way™ Kit
Includes:
One ruggedly designed, plastic worm-a-way™ vermicomposter with snap-on lid, manufactured from recycled materials one 162-page book, worms eat my garbage by mary appelhof, a biologist and well-known expert in worm composting, two easy-to-
Vermicomposter with easy-to-install vents, measures 19 x 16 x 12 in., worms eat my garbage book by mary appelhof, 162 pages, garden fork, assembly and set-up instructions
Description
Three great products work together to teach about the importance of composting and the role that worms play in it. For grades 4 to 8.
- Worm-a-Way™ Composting Kit (S8743) uses redworms to turn food waste into nutrient-rich humus that can be used for planting in the classroom
- Wormania! Video (S8745) teaches students about worms and their role in building soil (VHS, 26 min.)
- Worms Eat Our Garbage (S8746) activities book includes reproducible activities and teacher's guides, 232 pages
- Vermicomposter with easy-to-install vents; measures 19 × 16 × 12 in. (48.3 × 40.6 × 30.5cm) Worms Eat My Garbage book by Mary Appelhof, 162 pages
- Garden fork
- Assembly and set-up instructions
Specifications
Specifications
Product Type | Wormania! Video to Compliment Worm-A-Way™ Kit |
Grade | 4 to 8 |
For Use With (Application) | To teach students about worms and their role in building soil |
Includes | Vermicomposter with easy-to-install vents, measures 19 x 16 x 12 in., worms eat my garbage book by mary appelhof, 162 pages, garden fork, assembly and set-up instructions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction