Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worm Composting Kit and Video
SCI_ED

Catalog No. p-2369502
Click to view available options
Product Type:
Redworms Coupon
Wormania! Video to Compliment Worm-A-Way™ Kit
Includes:
One ruggedly designed, plastic worm-a-way™ vermicomposter with snap-on lid, manufactured from recycled materials one 162-page book, worms eat my garbage by mary appelhof, a biologist and well-known expert in worm composting, two easy-to-
Vermicomposter with easy-to-install vents, measures 19 x 16 x 12 in., worms eat my garbage book by mary appelhof, 162 pages, garden fork, assembly and set-up instructions

Composts 2 to 3 lb. of garbage each week.

Three great products work together to teach about the importance of composting and the role that worms play in it. For grades 4 to 8.

  • Worm-a-Way™ Composting Kit (S8743) uses redworms to turn food waste into nutrient-rich humus that can be used for planting in the classroom
  • Wormania! Video (S8745) teaches students about worms and their role in building soil (VHS, 26 min.)
  • Worms Eat Our Garbage (S8746) activities book includes reproducible activities and teacher's guides, 232 pages
Kit Includes:
  • Vermicomposter with easy-to-install vents; measures 19 × 16 × 12 in. (48.3 × 40.6 × 30.5cm) Worms Eat My Garbage book by Mary Appelhof, 162 pages
  • Garden fork
  • Assembly and set-up instructions

Specifications

Product Type Wormania! Video to Compliment Worm-A-Way™ Kit
Grade 4 to 8
For Use With (Application) To teach students about worms and their role in building soil
Includes Vermicomposter with easy-to-install vents, measures 19 x 16 x 12 in., worms eat my garbage book by mary appelhof, 162 pages, garden fork, assembly and set-up instructions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.