Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WQ 1
GREENER_CHOICESDP

Catalog No. 715650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-565-0 50 mg
71-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-565-0 Supplier Tocris Bioscience™ Supplier No. 7156/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective σ1 receptor antagonist; active in vivo

WQ 1 is a high affinity and selective σ1 receptor antagonist (pKi values are 10.28-10.85, 5.81-6.53 and 5.80 at σ1, σ2 and DAT). Displays σ1/σ2 selectivity ratio of 29512. Selectively blocks binge eating without a general inhibition of food intake in rats.

Specifications

Chemical Name or Material 2,3-Dihydro-2-[1-(phenylmethyl)-4-piperidinyl]-4 H-1-benzothiopyran-4-one
CAS 1204401-49-9
Quantity 50 mg
Target Sigma1 Receptor Antagonists
Molecular Formula C21H23NOS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.