Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WRAP53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555773 Supplier Invitrogen™ Supplier No. PA555773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGAVSVWDTD GPGNDGKPEP VLSFLPQKDC TNGVSLHPSL PLLATASGQR VFPEPTESGD EGEELGLPLL STRHVHLECR LQLWWCGGAP DSSIPD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WRAP53
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WRAP53
Gene Accession No. Q9BUR4
Gene Alias BC021790; DKCB3; TCAB1; Telomerase Cajal body protein 1; WD repeat containing antisense to TP53; WD repeat containing, antisense to TP53; WD repeat containing, antisense to Trp53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; Wdr79; WRAP53; WRAP53beta
Gene Symbols WRAP53
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WRAP53. Recombinant protein control fragment (Product #RP-93295).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.