Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WRAP53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110990
Description
Immunogen sequence: MKTLETQPLA PDCCPSDQDP APAHPSPHAS PMNKNADSEL MPPPPERGDP PRLSPDPVAG SAVSQELREG DPVSLSTPLE TEFGSPSELS PRIEEQELSE NT.
TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.
Specifications
| WRAP53 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUR4 | |
| WRAP53 | |
| Recombinant Protein corresonding to Human WRAP53. Recombinant protein control fragment (Product #RP-105817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| WRAP53 | |
| BC021790; DKCB3; TCAB1; Telomerase Cajal body protein 1; WD repeat containing antisense to TP53; WD repeat containing, antisense to TP53; WD repeat containing, antisense to Trp53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; Wdr79; WRAP53; WRAP53beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction