Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WRAP53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120692 Supplier Invitrogen™ Supplier No. PA5120692
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse kidney. Immunogen sequence: AVSQELREGD PVSLSTPLET EFGSPSELSP RIEEQELSEN TSLPAEEANG SLSEEEANGP ELGSGKAMED TSGEPAAEDE GDTAWNYSFS QLPRFLSGS.

TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WRAP53
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene WRAP53
Gene Accession No. Q8VC51
Gene Alias BC021790; DKCB3; TCAB1; Telomerase Cajal body protein 1; WD repeat containing antisense to TP53; WD repeat containing, antisense to TP53; WD repeat containing, antisense to Trp53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; Wdr79; WRAP53; WRAP53beta
Gene Symbols WRAP53
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 62-160 of human WRAP53 (NP_060551.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216853
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.